Learn More
Sino Biological Human CD69 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11150T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD69 (rh CD69; Catalog#11150-H08H; NP_001772.1; Ser62-Lys199). CD69 specific IgG was purified by Human CD69 affinity chromatography.
Catalog No. 50-237-1784