Learn More
ACROBiosystems Human CD7 200ug
Catalog No. 502535341
Encompass_Preferred
Alexa Fluor 488-Labeled Human CD7 Protein His Tag (CD7-HA2H9) is produced via conjugation of AF488 to Human CD7 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).
Catalog No. 50-253-5341 Supplier ACROBiosystems Supplier No. CD7HA2H9200UG
Alexa Fluor 488-Labeled Human CD7 Protein His Tag (CD7-HA2H9) is produced via conjugation of AF488 to Human CD7 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).