Learn More
ACROBiosystems Human CD7 25tests
Catalog No. 502535342
Encompass_Preferred
APC-Labeled Human CD7 Protein His Tag (CD7-HA2H6) is produced via conjugation of APC to Human CD7 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).
Catalog No. 50-253-5342 Supplier ACROBiosystems Supplier No. CD7HA2H625TESTS
APC-Labeled Human CD7 Protein His Tag (CD7-HA2H6) is produced via conjugation of APC to Human CD7 Protein His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).