Learn More
ACROBiosystems PE-Labeled Human CD7 Protein, His TagStar Staining
Catalog No. 502331986
Encompass_Preferred
PE-Labeled Human CD7 Protein, His Tag (CD7-HP2H6) is produced via conjugation of PE to Human CD7 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).
Catalog No. 50-233-1986 Supplier ACROBiosystems Supplier No. CD7HP2H625TESTS
PE-Labeled Human CD7 Protein, His Tag (CD7-HP2H6) is produced via conjugation of PE to Human CD7 Protein, His Tag with a new generation site-specific technology under Star Staining labeling platform. Human CD7 Protein, His Tag is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).