Learn More
Sino Biological Anti-CD7 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 11028MM21C100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Human CD7 Plasmid. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8992