Learn More
Sino Biological Anti-CD7 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 11028MM12F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD7 (rh CD7; Catalog#11028-H08H; NP_006128.1; Met1-Pro180) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-1872