Learn More
Sino Biological Human CD7 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 11028MM21P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with Human CD7 Plasmid. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2056