Learn More
Sino Biological Human CD7 MAb 25TS (Sino Biological)
Supplier:  Sino Biological 11028MM14P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD7 (rh CD7; Catalog#11028-H08H; NP_006128.1; Met1-Pro180) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-237-2019