Learn More
Sino Biological Human CD7 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 11028H02H1
Encompass_Preferred
A DNA sequence encoding the human CD7 (NP_006128.1) (Met1-Pro180) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-2242