Learn More
ACROBiosystems ACROBiosystems Human CD7 Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CD7H52531MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD7, Fc Tag (CD7-H5253) is expressed from human 293 cells (HEK293). It contains AA Ala 26 - Pro 180 (Accession # P09564-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 42.9 kDa. The protein migrates as 55-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human CD7, Fc Tag (Cat. No. CD7-H5253) at 1 μg/mL (100 μL/well) can bind Anti-CD7 antibody, Mouse IgG1 with a linear range of 1-2 ng/mL (QC tested).
Catalog No. 50-210-6208