Learn More
Sino Biological Recombinant Human CD70/CD27 Ligand Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10780H01H51
Encompass_Preferred
A DNA sequence encoding the Human CD70(NP_001243.1) (Gln39-Pro193) was expressed with the Fc region of human IgG1 at the N-terminus. This construct has a trimer design.
Catalog No. 50-241-1713