Learn More
ACROBiosystems Human CD72, Mouse IgG2a Fc Tag (CD2-H5255) is expressed from human 293 cells (HEK293). It contains AA Arg 117 - Asp 359 (Accession # P21855-1).
Catalog No. 502231368
Encompass_Preferred
B-cell differentiation antigen CD72 is also known as Lyb-2. CD72 contains one C-type lectin domain. CD72 is a protein active in the immune system of animals. CD72 consists of two identical halves, each of about 39-43 kD, and is a C-type lectin. Its primarily locus of expression is B-cells, where it appears to mediate aspects of B-cell - T-cell interaction. CD72 plays a role in B-cell proliferation and differentiation. CD72 is a ligand for CD5.
This item is not returnable. View return policy
Catalog No. 50-223-1368 Supplier ACROBiosystems Supplier No. CD2H5255100UG
B-cell differentiation antigen CD72 is also known as Lyb-2. CD72 contains one C-type lectin domain. CD72 is a protein active in the immune system of animals. CD72 consists of two identical halves, each of about 39-43 kD, and is a C-type lectin. Its primarily locus of expression is B-cells, where it appears to mediate aspects of B-cell - T-cell interaction. CD72 plays a role in B-cell proliferation and differentiation. CD72 is a ligand for CD5.