Learn More
Sino Biological Human CD72 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 12507H08H1
Encompass_Preferred
A DNA sequence encoding the human CD72 (NP_001773.1) (Arg117-Asp359) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2406