Learn More
Bon Opus Biosciences Recombinant Human CD74 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP103950UG
Encompass_Preferred
Recombinant Human CD74 (C-His) is produced by Human Cells. The target gene encoding Gln73-Met232 is expressed with a C-His tag. Accession# NP_004346.1. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1039
Catalog No. 50-217-2638