Learn More
ACROBiosystems ACROBiosystems Human CD74 / DHLAG Protein, Fc Tag (MALS verified)
Supplier:  ACROBiosystems CD4H5256100UG
Encompass_Preferred
Recombinant Protein;100UG;Human CD74, Fc Tag (CD4-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 73 - Met 232 (Accession # P04233-2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 47.9 kDa. The protein migrates as 60-65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7236