Learn More
Sino Biological Anti-CD74 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11091MM01P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD74 (rh CD74; Catalog#11091-H07H; NP_004346.1; Gln73-Met232) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4835