Learn More
Sino Biological Anti-CD74 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11091T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD74 (rh CD74; Catalog#11091-H07H; NP_004346.1; Gln73-Met232). CD74 specific IgG was purified by Human CD74 affinity chromatography.
Catalog No. 50-255-4832