Learn More
ACROBiosystems Human CD79A&CD79B Heterodimer Protein, Mouse IgG2a Fc,Flag Tag&Mouse IgG2a Fc,His Tag (CDB-H52W3) is expressed from human 293 cells (HEK293). It contains AA Leu 33 - Arg 143 (CD79A) & Ala 29 - Asp 159 (CD79B)
Catalog No. 502230577
Encompass_Preferred
CD79a and CD79b heterodimers are the important signaling components of B cell receptor (BCR) complex which plays a crucial role in B cell development and antibody production. BCR complexes are composed of a ligand-binding receptor (membrane immunoglobulin; mIg), non-covalently associated with the signaling com-ponent, a disulfide-linked heterodimer of CD79a (Igα) and CD79b (Igβ). Both CD79 subunits consist of an Ig-like domain, a transmembrane (TM) region, and a longer cytoplasmic tail (CYT) containing an immune-receptor tyrosine-based activation motif (ITAM).
Catalog No. 50-223-0577 Supplier ACROBiosystems Supplier No. CDBH52W3100UG
CD79a and CD79b heterodimers are the important signaling components of B cell receptor (BCR) complex which plays a crucial role in B cell development and antibody production. BCR complexes are composed of a ligand-binding receptor (membrane immunoglobulin; mIg), non-covalently associated with the signaling com-ponent, a disulfide-linked heterodimer of CD79a (Igα) and CD79b (Igβ). Both CD79 subunits consist of an Ig-like domain, a transmembrane (TM) region, and a longer cytoplasmic tail (CYT) containing an immune-receptor tyrosine-based activation motif (ITAM).