Learn More
ACROBiosystems Human CD8 Alpha and Beta Heterodimer Protein, Low Endotoxin, His Tag and Tag Free, 1 mg
Catalog No. 502719786
Encompass_Preferred
Human CD8 alpha&beta heterodimer protein is an integral membrane glycoprotein that plays an essential role in the immune response. It is expressed from human HEK293 cells and contains amino acids Ser 22 to Asp 182 and Leu 22 to Pro 170. This protein is supplied as a lyophilized powder with a His tag and tag-free configuration, featuring a purity of >95% as determined by SDS-PAGE. The protein migrates as 28-30 kDa and 31-33 kDa under reducing conditions due to glycosylation.
  • Expressed in HEK293 cells
  • Low endotoxin level of <0.01 EU per μg
  • Purity >95% by SDS-PAGE
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose
  • Binding activity confirmed via ELISA and BLI (affinity constant of 66.8 nM)
  • Stable at -20°C to -70°C for 12 months in lyophilized state
Catalog No. 50-271-9786 Supplier ACROBiosystems Supplier No. CDAH52W51MG
Human CD8 alpha&beta heterodimer protein is an integral membrane glycoprotein that plays an essential role in the immune response. It is expressed from human HEK293 cells and contains amino acids Ser 22 to Asp 182 and Leu 22 to Pro 170. This protein is supplied as a lyophilized powder with a His tag and tag-free configuration, featuring a purity of >95% as determined by SDS-PAGE. The protein migrates as 28-30 kDa and 31-33 kDa under reducing conditions due to glycosylation.
  • Expressed in HEK293 cells
  • Low endotoxin level of <0.01 EU per μg
  • Purity >95% by SDS-PAGE
  • Lyophilized from 0.22 μm filtered PBS, pH 7.4 with trehalose
  • Binding activity confirmed via ELISA and BLI (affinity constant of 66.8 nM)
  • Stable at -20°C to -70°C for 12 months in lyophilized state