Learn More
ACROBiosystems Human CD8 alpha beta 200ug
Catalog No. 502535324
Encompass_Preferred
Biotinylated Human CD8ACD8B Heterodimer Protein HisAvitagTag Free (CDA-H82W7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (CD8A) Leu 22 - Pro 170 (CD8B) (Accession # P01732-1 (CD8A) P10966-1 (CD8B) ).
Catalog No. 50-253-5324 Supplier ACROBiosystems Supplier No. CDAH82W7200UG
Biotinylated Human CD8ACD8B Heterodimer Protein HisAvitagTag Free (CDA-H82W7) is expressed from human 293 cells (HEK293). It contains AA Ser 22 - Asp 182 (CD8A) Leu 22 - Pro 170 (CD8B) (Accession # P01732-1 (CD8A) P10966-1 (CD8B) ).