Learn More
Sino Biological Human CD8 alpha Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000785534
Encompass_Preferred
The human CD8A ELISA Pair Set is for the quantitative determination of human CD8A.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD8Acoated on a 96-well plate. Standards and samples are added to the wells, andany CD8A present binds to the immobilized antibody. The wells are washed and ahorseradish peroxidase conjugated mouse anti-CD8A monoclonal antibody is thenadded, producing an antibody-antigen-antibody "sandwich". The wells are againwashed and TMB substrate solution is loaded, which produces color in proportionto the amount of CD8A present in the sample. To end the enzyme reaction, thestop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85534 Supplier Sino Biological Supplier No. SEK109805
The human CD8A ELISA Pair Set is for the quantitative determination of human CD8A.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for CD8Acoated on a 96-well plate. Standards and samples are added to the wells, andany CD8A present binds to the immobilized antibody. The wells are washed and ahorseradish peroxidase conjugated mouse anti-CD8A monoclonal antibody is thenadded, producing an antibody-antigen-antibody "sandwich". The wells are againwashed and TMB substrate solution is loaded, which produces color in proportionto the amount of CD8A present in the sample. To end the enzyme reaction, thestop solution is added and absorbances of the microwell are read at 450 nm.