Learn More
Raybiotech Inc Human CD80 (B7-1) ELISA Kit
Supplier:  Raybiotech Inc ELHCD801
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 12 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human B7-1 / CD80.
Catalog No. 50-194-9102