Learn More
Sino Biological Recombinant Anti-B7-1/CD80 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10698R77520
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD80 / B7-1 (rh CD80 / B7-1; Catalog#10698-H08H; NP_005182.1; Met1-Asn242).
Catalog No. 50-255-5203