Learn More
American Research Products Inc Human CD81 antigen CD81 ELIS
Catalog No. 5000327281
Encompass
Human CD81 antigen (CD81) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27281 Supplier American Research Products Inc Supplier No. CSBEL004960HU
Human CD81 antigen (CD81) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich