Learn More
Sino Biological Anti-CD81 Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 14244MM03A100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD81/TAPA1 (rh CD81/TAPA1; Catalog#14244-HNCH; NP_004347.1; Phe113-Lys201) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-240-9910