Learn More
Sino Biological Anti-CD81 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 14244MM05H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD81 (rh CD81; Catalog#14244-HNCH; NP_004347.1; Phe113-Lys201). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8433