Learn More
Sino Biological Anti-CD81 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14244RP0120
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD81 / TAPA1 (rh CD81 / TAPA1; Catalog#14244-HNCH; NP_004347.1; Phe113-Lys201). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4142