Learn More
Sino Biological Anti-CD83 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 11763MM02P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD83 (rh CD83; Catalog#11763-H08H; Q01151-1; Met1-Ala143) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4575