Learn More
Sino Biological Recombinant Anti-CD83 Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 11763R105P100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD83 (rh CD83; Catalog#11763-H08H; Q01151-1; Met1-Ala143) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-255-4564