Learn More
Raybiotech Inc Human CD86 (B7-2) ELISA Kit
Supplier:  Raybiotech Inc ELHCD861
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.82 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human CD86 (B7-2)
Catalog No. 50-194-9104