Learn More
Sino Biological Recombinant Anti-B7-2/CD86 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10699R11820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD86 / B7-2 (rh CD86; Catalog#10699-H08H; NP_008820.2; Met 1-His 239).
Catalog No. 50-241-1742