Learn More
Sino Biological Anti-B7-2/CD86 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10699T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD86 (rh CD86; Catalog#10699-H08H; NP_008820.2; Met1-His239). CD86 specific IgG was purified by Human CD86 affinity chromatography.
Catalog No. 50-255-5197