Learn More
Sino Biological Recombinant Human B7-2/CD86 Protein (hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10699H41HB100
Encompass_Preferred
A DNA sequence encoding the Human CD86 (NP_008820.4) (Met1-His239) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-5199