Learn More
ACROBiosystems ACROBiosystems Human CD89 / FCAR Protein, His Tag
Supplier:  ACROBiosystems CD9H52HA1MG
Encompass_Preferred
Recombinant Protein;1MG;Human CD89, His Tag (CD9-H52Ha) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 227 (Accession # P24071-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 25.4 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6501