Learn More
Sino Biological Recombinant Human CD8 alpha Protein (ECD, His Tag), Amine-Labeled Biotinylated 100
Supplier:  Sino Biological 10980H08SB100
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD8A (NP_001759.3) (Met1-Asp182) was expressed with a C-terminal polyhistidine tag. The purified protein was biotinylated in vitro.
Catalog No. 50-222-3367