Learn More
Sino Biological Recombinant Human CD8 alpha Protein (ECD, His Tag), HPLC-verified
Supplier:  Sino Biological 10980H08S500
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human CD8A (NP_001759.3) (Met1-Asp182) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-222-3365