Learn More
BIOHIPPO Human CD8b Microsample
SDP
Catalog No. 5000314867
Encompass
Human CD8b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD8b in Tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 56 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD8b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD8b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD8b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14867 Supplier BIOHIPPO Supplier No. BHE15210828
Human CD8b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CD8b in Tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 56 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CD8b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CD8b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CD8b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color