Learn More
Sino Biological Recombinant Human CD9 Protein (Nanodisc, Full-length, His-GFP Tag), Fluorescent
Catalog No. 502992923
Encompass_Preferred
A DNA sequence encoding the Human CD9 (P21926) (Met1-Val228) was expressed with a N-terminal polyhistidine tag followed by a GFP Tag. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-299-2923 Supplier Sino Biological Supplier No. 11029H87HNA20
A DNA sequence encoding the Human CD9 (P21926) (Met1-Val228) was expressed with a N-terminal polyhistidine tag followed by a GFP Tag. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.