Learn More
Sino Biological Anti-CD9 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 11029MM01C25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD9 (rh CD9; Catalog#11029-H08H; NP_001760.1; Ser 112-Ile 195) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-255-4967