Learn More
Sino Biological Human CD9-VLP (Full Length) Protein (Flag Tag)
Supplier:  Sino Biological 11029H25H100
Encompass_Preferred
A DNA sequence encoding the Human CD9-VLP (Full Length) (P21926) (Met1-Val228) was fused with a signal peptide at the N-terminus and a flag tag at the C-terminus.
Catalog No. 50-255-4973