Learn More
Sino Biological Anti-C1qR1/CD93 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 12589MM0420
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD93/C1qR (rh CD93/C1qR; Catalog#12589-H08H; Q9NPY3; Met1-Lys580). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8111