Learn More
Sino Biological Anti-C1qR1/CD93 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 12589RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD93 / C1qR (rh CD93 / C1qR; Catalog#12589-H08H; Q9NPY3; Met1-Lys580). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2136