Learn More
Sino Biological Recombinant Human CD96 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 11202H08HB20
Encompass_Preferred
A DNA sequence encoding the human CD96 isoform 2 (P40200-2) extracellular domain (Met1-Met503) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-4792