Learn More
Sino Biological Recombinant Human CD98 Protein (ECD, His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 12206H07HB100
Encompass_Preferred
A DNA sequence encoding the human SLC3A2 (P08195-1) (Arg 206-Ala 630) was expressed, with a polyhistidine tag at the N-terminus.The purified protein was biotinylated in vitro.
Catalog No. 50-228-2411