Learn More
Raybiotech Inc Human CD99 ELISA Kit
Supplier:  Raybiotech Inc ELHCD991
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.062 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human CD99.
Catalog No. 50-194-9105