Learn More
Sino Biological Human CDC2 & CCNE1 Protein 50ug (Sino Biological)
Supplier:  Sino Biological CT027H0907B50
Encompass_Preferred
A DNA sequence encoding the human CDC2 (NP_001777.1) (Met1-Met297) was expressed with a GST tag at the N-terminus, constructed the plasmid 1; A DNA sequence encoding the human CCNE1 (NP_001229.1) (Met1-Ala410) was expressed with a polyhistidine tag at the N-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterotrimer was purified.
Catalog No. 50-237-1986