Learn More
Sino Biological Anti-Cdc25B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105166T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Cdc25B, and purified by antigen affinity chromatography.
Catalog No. 50-240-8850