Learn More
Sino Biological Anti-CDC42 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10062RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CDC42 (rh CDC42; Catalog#10062-H09E; P60953-2; Met1-Cys188). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-1616