Learn More
Sino Biological Anti-CDCP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 203663T40100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human CDCP1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-255-3909