Learn More
Sino Biological Human CDH1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10204MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Cadherin-1 / CDH1 (rh Cadherin-1; Catalog#10204-H08H; Met 1-Ile 707; NP_004351.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2090